
3,5-Bis(trifluoromethyl)-N-(bromoacetyl)aniline 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:99468-72-1
:
Formula:C₁₀H₆BrF₆NO
MDL Number:MFCD00662315
:350.06 g/mol
Melting Pt:100…101 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together