2-Chloro-3,6-difluorophenylacetonitrile 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:261762-54-3
:
Formula:C₈H₄ClF₂N
MDL Number:MFCD01631447
ADR:6.1,III
:187.58 g/mol
Storage Temperature:Ambient
UN:2811
Frequently Bought Together