

Diethyl bromomalonate 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:685-87-0
:
Formula:C₇H₁₁BrO₄
MDL Number:MFCD00009138
:239.07 g/mol
Boiling Pt:140…141 °C/30mmHg
Density:1.402 g/cm³ (25 °C)
Flash Pt:111 °C
Storage Temperature:Ambient
EINECS:211-683-1
UN:3265
ADR:8,II
:GHS05
:GHS07
Frequently Bought Together