Order Entry
Czech Republic
ContactUsLinkComponent
Ethyl-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate 98%
Irritant
Ethyl-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate 98%
Catalog # APOSOR9928-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Ethyl-5-formyl-2,4-dimethyl-1H-pyrrole-3-carboxylate 98%
Catalog # APOSOR9928-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :195.22 g/mol
Melting Pt:165…170 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:2199-59-9
Formula:C₁₀H₁₃NO₃
MDL Number:MFCD00030352