Order Entry
Czech Republic
ContactUsLinkComponent
3-[(Dimethylamino)methyl]-4-hydroxy-5-methoxybenzenecarbaldehyde 95%
Irritant
3-[(Dimethylamino)methyl]-4-hydroxy-5-methoxybenzenecarbaldehyde 95%
Catalog # APOSOR9894-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
3-[(Dimethylamino)methyl]-4-hydroxy-5-methoxybenzenecarbaldehyde 95%
Catalog # APOSOR9894-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :209.25 g/mol
Melting Pt:140…141 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:87743-10-0
Formula:C₁₁H₁₅NO₃
MDL Number:MFCD03011572