
4-(3-Bromophenyl)-2-methylpyrimidine 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:844891-12-9
:
Formula:C₁₁H₉BrN₂
MDL Number:MFCD06659074
:249.12 g/mol
Melting Pt:66…69 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together