Order Entry
Czech Republic
ContactUsLinkComponent
 
Irritant
Imidazo[1,2-a]pyridine-3-carboxylic acid 98%
Catalog # APOSOR9691-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Imidazo[1,2-a]pyridine-3-carboxylic acid 98%
Catalog # APOSOR9691-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :162.1454 g/mol
Melting Pt:195.5…197 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:6200-60-8
Formula:C₈H₆N₂O₂
MDL Number:MFCD01720458