Order Entry
Czech Republic
ContactUsLinkComponent
2-Oxo-2,5,6,7-tetrahydro-1H-[1]pyrindine-3-carboxylic acid 95%
2-Oxo-2,5,6,7-tetrahydro-1H-[1]pyrindine-3-carboxylic acid 95%
Catalog # APOSOR965109-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Oxo-2,5,6,7-tetrahydro-1H-[1]pyrindine-3-carboxylic acid 95%
Catalog # APOSOR965109-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :179.18 g/mol
Storage Temperature:Ambient
CAS Number:115122-63-9
Formula:C₉H₉NO₃
MDL Number:MFCD07801152