2-Cyclopropylpyrimidine-5-carboxylic acid 95%
Catalog # APOSOR963924-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:164.160 g/mol
Storage Temperature:Ambient
CAS Number:648423-79-4
Formula:C₈H₈N₂O₂
MDL Number:MFCD09054852