3-Methylpiperidine-1,3-dicarboxylic acid-1-tert-butyl ester 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:534602-47-6
:
Formula:C₁₂H₂₁NO₄
MDL Number:MFCD08461329
:243.31 g/mol
Storage Temperature:Ambient
Frequently Bought Together