Order Entry
Czech Republic
ContactUsLinkComponent
(1-Phenyl-1H-pyrazol-4-yl)methanol 97%
(1-Phenyl-1H-pyrazol-4-yl)methanol 97%
Catalog # APOSOR953502-1G
Supplier:  Apollo Scientific
CAS Number:  
(1-Phenyl-1H-pyrazol-4-yl)methanol 97%
Catalog # APOSOR953502-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :174.2 g/mol
Storage Temperature:Ambient
CAS Number:70817-26-4
Formula:C₁₀H₁₀N₂O
MDL Number:MFCD00159595