2,3-Dihydrobenzo[1,4]dioxine-2-carboxylic acid methyl ester 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3663-79-4
:
Formula:C₁₀H₁₀O₄
MDL Number:MFCD00297092
:194.19 g/mol
Storage Temperature:Ambient
Frequently Bought Together