Methyl pyrazolo[1,5-a]pyridine-2-carboxylate 95%
Catalog # APOSOR953065-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
Storage Temperature:Ambient
CAS Number:151831-21-9
Formula:C₉H₈N₂O₂
MDL Number:MFCD07367039