Order Entry
Czech Republic
ContactUsLinkComponent
2-Methoxyethylsulphonyl chloride 95%
2-Methoxyethylsulphonyl chloride 95%
Catalog # APOSOR952816-5G
Supplier:  Apollo Scientific
CAS Number:  
2-Methoxyethylsulphonyl chloride 95%
Catalog # APOSOR952816-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :158.6 g/mol
Storage Temperature:Ambient
CAS Number:51517-01-2
Formula:C₃H₇ClO₃S
MDL Number:MFCD10032391