
2-Methylimidazo[1,2-a]pyridine-3-carbohydrazide
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:144835-67-6
:
Formula:C₉H₁₀N₄O
MDL Number:MFCD00269303
Melting Pt:180…185 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together