Order Entry
Czech Republic
ContactUsLinkComponent
4,6-Dichloro-5-nitropyrimidine 97%
Irritant
4,6-Dichloro-5-nitropyrimidine 97%
Catalog # APOSOR9521-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
4,6-Dichloro-5-nitropyrimidine 97%
Catalog # APOSOR9521-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :193.98 g/mol
Melting Pt:100…103 °C
EINECS:224-340-6
 :GHS07
Storage Temperature:Refrigerator
CAS Number:4316-93-2
Formula:C₄HCl₂N₃O₂
MDL Number:MFCD00006107