

2,6-Dinitro-4-methylanisole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:29455-11-6
:
Formula:C₈H₈N₂O₅
MDL Number:MFCD00192097
Melting Pt:124 °C
UN:2930
ADR:6.1+4.1,II
:GHS02
:GHS06
Storage Temperature:Ambient
Frequently Bought Together