Order Entry
Czech Republic
ContactUsLinkComponent
2-(4-Methylphenoxy)ethylamine 95%
2-(4-Methylphenoxy)ethylamine 95%
Catalog # APOSOR951144-1G
Supplier:  Apollo Scientific
CAS Number:  
2-(4-Methylphenoxy)ethylamine 95%
Catalog # APOSOR951144-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :151.20 g/mol
Storage Temperature:Ambient
CAS Number:26583-58-4
Formula:C₉H₁₃NO
MDL Number:MFCD02598961