Order Entry
Czech Republic
ContactUsLinkComponent
2-Chloro-3,3-diethoxyprop-1-ene 95%
2-Chloro-3,3-diethoxyprop-1-ene 95%
Catalog # APOSOR950581-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-3,3-diethoxyprop-1-ene 95%
Catalog # APOSOR950581-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :164.63 g/mol
Storage Temperature:Ambient
CAS Number:7575-33-9
Formula:C₇H₁₃ClO₂
MDL Number:MFCD23102091