(±)-2,6,10,10-Tetramethyl-1-oxaspiro[4.5]-5-decene 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:36431-72-8
:
Formula:C₁₃H₂₂O
MDL Number:MFCD00085214
:194.32 g/mol
Storage Temperature:Ambient
Frequently Bought Together