Methyl tetrahydro-4-oxo-2H-thiopyran-3-carboxylate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:4160-61-6
:
Formula:C₇H₁₀O₃S
MDL Number:MFCD00203485
:174.214 g/mol
Storage Temperature:Ambient
Frequently Bought Together