Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Expresní objednávka
Stav objednávky
Váš účet
Košík
Czech Republic
Expresní objednávka
Stav objednávky
Váš účet
Košík
ContactUsLinkComponent
Home
4-(Propionylamino)benzoic acid
4-(Propionylamino)benzoic acid 95%
4-(Propionylamino)benzoic acid 95%
4-(Propionylamino)benzoic acid 95%
APOSOR945554-1G
:
Apollo Scientific
:
4-(Propionylamino)benzoic acid 95%
APOSOR945554-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
193.2 g/mol
:
Ambient
:
19313-85-0
:
C₁₀H₁₁NO₃
:
MFCD01365922