
3-Phenyl-1H-pyrazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2458-26-6
:
Formula:C₉H₈N₂
MDL Number:MFCD00159654
:144.18 g/mol
Boiling Pt:314 °C
Melting Pt:75…82 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together