Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl-3,3-dimethoxy-2-methylpropanoate
Methyl-3,3-dimethoxy-2-methylpropanoate 90%
Methyl-3,3-dimethoxy-2-methylpropanoate 90%
APOSOR944664-1G
:
Apollo Scientific
:
Methyl-3,3-dimethoxy-2-methylpropanoate 90%
APOSOR944664-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
162.19 g/mol
:
Ambient
:
76526-43-7
:
C₇H₁₄O₄
:
MFCD19103373