Order Entry
Czech Republic
ContactUsLinkComponent
2-[[tert-Butyl(dimethyl)silyl]oxy]ethylamine 95%
2-[[tert-Butyl(dimethyl)silyl]oxy]ethylamine 95%
Catalog # APOSOR944134-1G
Supplier:  Apollo Scientific
CAS Number:  
2-[[tert-Butyl(dimethyl)silyl]oxy]ethylamine 95%
Catalog # APOSOR944134-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :175.35 g/mol
Storage Temperature:Ambient
CAS Number:101711-55-1
Formula:C₈H₂₁NOSi
MDL Number:MFCD18206132