Order Entry
Czech Republic
ContactUsLinkComponent
Imidazo[1,2-a]pyridine-3-carbonitrile 95%
Imidazo[1,2-a]pyridine-3-carbonitrile 95%
Catalog # APOSOR942541-1G
Supplier:  Apollo Scientific
CAS Number:  
Imidazo[1,2-a]pyridine-3-carbonitrile 95%
Catalog # APOSOR942541-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :143.15 g/mol
Storage Temperature:Ambient
CAS Number:6200-59-5
Formula:C₈H₅N₃
MDL Number:MFCD01720448