3-(Piperidin-1-ylmethyl)benzoic acid 97%
Catalog # APOSOR942220-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:219.29 g/mol
Storage Temperature:Ambient
CAS Number:158861-24-6
Formula:C₁₃H₁₇NO₂
MDL Number:MFCD05841589