Order Entry
Czech Republic
ContactUsLinkComponent
2-Phenyl-2-(2-pyridyl)acetonitrile 95%
2-Phenyl-2-(2-pyridyl)acetonitrile 95%
Catalog # APOSOR941974-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Phenyl-2-(2-pyridyl)acetonitrile 95%
Catalog # APOSOR941974-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :194.24 g/mol
Storage Temperature:Ambient
CAS Number:5005-36-7
Formula:C₁₃H₁₀N₂
MDL Number:MFCD00023497