Order Entry
Czech Republic
ContactUsLinkComponent
N,N,2,2-Tetramethyl-1,3-propanediamine 95%
N,N,2,2-Tetramethyl-1,3-propanediamine 95%
Catalog # APOSOR941777-100G
Supplier:  Apollo Scientific
CAS Number:  
N,N,2,2-Tetramethyl-1,3-propanediamine 95%
Catalog # APOSOR941777-100G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :130.24 g/mol
Storage Temperature:Ambient
CAS Number:53369-71-4
Formula:C₇H₁₈N₂
MDL Number:MFCD00044018