
N,N-Diethyl 3-borono-4-methylbenzenesulphonamide 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:871329-80-5
:
Formula:C₁₁H₁₈BNO₄S
MDL Number:MFCD08056356
:271.14 g/mol
Melting Pt:146…152 °C
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together