Order Entry
Czech Republic
ContactUsLinkComponent
6-Methoxy-4-chromanone 95%
6-Methoxy-4-chromanone 95%
Catalog # APOSOR939121-250MG
Supplier:  Apollo Scientific
CAS Number:  
6-Methoxy-4-chromanone 95%
Catalog # APOSOR939121-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :178.19 g/mol
Storage Temperature:Ambient
CAS Number:5802-17-5
Formula:C₁₀H₁₀O₃
MDL Number:MFCD00190159