Order Entry
Czech Republic
ContactUsLinkComponent
2-(Hydroxymethyl)-4-nitrobenzenol 98%
Irritant
2-(Hydroxymethyl)-4-nitrobenzenol 98%
Catalog # APOSOR938939-25G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2-(Hydroxymethyl)-4-nitrobenzenol 98%
Catalog # APOSOR938939-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :169.14 g/mol
Melting Pt:125 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:39224-61-8
Formula:C₇H₇NO₄
MDL Number:MFCD01321361