Order Entry
Czech Republic
ContactUsLinkComponent
2,2,6-Trimethyl-6-vinyltetrahydropyran-3-ol (mixture of isomers) 95%
2,2,6-Trimethyl-6-vinyltetrahydropyran-3-ol (mixture of isomers) 95%
Catalog # APOSOR938663-5G
Supplier:  Apollo Scientific
CAS Number:  
2,2,6-Trimethyl-6-vinyltetrahydropyran-3-ol (mixture of isomers) 95%
Catalog # APOSOR938663-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :170.25 g/mol
Storage Temperature:Ambient
CAS Number:14049-11-7
Formula:C₁₀H₁₈O₂
MDL Number:MFCD06200770