2-(Dimethylamino)propan-1-ol 95%
Catalog # APOSOR938625-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:103.17 g/mol
Storage Temperature:Ambient
CAS Number:15521-18-3
Formula:C₅H₁₃NO
MDL Number:MFCD00014425