Order Entry
Czech Republic
ContactUsLinkComponent
O-Acetyl-N-tert-butylhydroxylamine hydrochloride 95%
O-Acetyl-N-tert-butylhydroxylamine hydrochloride 95%
Catalog # APOSOR938608-1G
Supplier:  Apollo Scientific
CAS Number:  
O-Acetyl-N-tert-butylhydroxylamine hydrochloride 95%
Catalog # APOSOR938608-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :167.63 g/mol
Storage Temperature:Ambient
CAS Number:851074-40-3
Formula:C₆H₁₃NO₂
MDL Number:MFCD12545957