Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(4-Morpholinylcarbonyl)phenylboronic acid
2-(4-Morpholinylcarbonyl)phenylboronic acid 95%
2-(4-Morpholinylcarbonyl)phenylboronic acid 95%
2-(4-Morpholinylcarbonyl)phenylboronic acid 95%
APOSOR9384-1G
:
Apollo Scientific
:
2-(4-Morpholinylcarbonyl)phenylboronic acid 95%
APOSOR9384-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
150…152 °C
:
GHS07
:
Refrigerator
:
874219-17-7
:
C₁₁H₁₄BNO₄
:
MFCD08235036