
Dicyclohexyl(2',4',6'-triisopropyl-3,6-dimethoxy-[1,1'-biphenyl]-2-yl)phosphine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1070663-78-3
:
Formula:C₃₅H₅₃O₂P
MDL Number:MFCD11973797
Melting Pt:187…195 °C
:536.78 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together