Order Entry
Czech Republic
ContactUsLinkComponent
 
N-(n-Propyl)-2-(2-pyridyl)ethylamine 97%
Catalog # APOSOR938116-1G
Supplier:  Apollo Scientific
CAS Number:  
N-(n-Propyl)-2-(2-pyridyl)ethylamine 97%
Catalog # APOSOR938116-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:55496-57-6
Formula:C₁₀H₁₆N₂
MDL Number:MFCD00023544