Order Entry
Czech Republic
ContactUsLinkComponent
[2-(2-Methoxyphenoxy)ethyl]methylamine 97%
[2-(2-Methoxyphenoxy)ethyl]methylamine 97%
Catalog # APOSOR937878-1G
Supplier:  Apollo Scientific
CAS Number:  
[2-(2-Methoxyphenoxy)ethyl]methylamine 97%
Catalog # APOSOR937878-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :181.24 g/mol
Storage Temperature:Ambient
CAS Number:72955-82-9
Formula:C₁₀H₁₅NO₂
MDL Number:MFCD07400597