2,7-Dimethoxy-1,4,5,8-tetrahydronaphthalene 97%
Catalog # APOSOR937133-5G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Typ balení:Bottle
- Bal.:5 g
- Sekvence:3173
Specifications
About this item
:
:192.26 g/mol
Storage Temperature:Ambient
CAS Number:1614-82-0
Formula:C₁₂H₁₆O₂
MDL Number:MFCD00269695