
4-Iodobenzylamine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:59528-27-7
:
Formula:C₇H₉ClIN
MDL Number:MFCD04039324
:269.51 g/mol
Melting Pt:275…280 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together