Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-(2-Methoxyethyl)ethylamine
N-(2-Methoxyethyl)ethylamine 97%
N-(2-Methoxyethyl)ethylamine 97%
N-(2-Methoxyethyl)ethylamine 97%
APOSOR936987-5G
:
Apollo Scientific
:
N-(2-Methoxyethyl)ethylamine 97%
APOSOR936987-5G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
5 g
Seq:
3173
:
:
103.17 g/mol
:
Ambient
:
34322-82-2
:
C₅H₁₃NO
:
MFCD00144876