Order Entry
Czech Republic
ContactUsLinkComponent
5-Ethoxy-2-mercaptobenzimidazole 97%
5-Ethoxy-2-mercaptobenzimidazole 97%
Catalog # APOSOR936925-25G
Supplier:  Apollo Scientific
CAS Number:  
5-Ethoxy-2-mercaptobenzimidazole 97%
Catalog # APOSOR936925-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :194.25 g/mol
Storage Temperature:Ambient
CAS Number:55489-15-1
Formula:C₉H₁₀N₂OS
MDL Number:MFCD01326487