

1,3-Divinyl-1,1,3,3-tetramethyldisilazane 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:7691-02-3
:
Formula:C₈H₁₉NSi₂
MDL Number:MFCD00042846
:185.42 g/mol
Boiling Pt:160 °C
Density:0.82 g/cm³ (25 °C)
Flash Pt:42 °C
Storage Temperature:Ambient
UN:2924
ADR:3+8,III
:GHS02
:GHS06
Frequently Bought Together