
1-(4-Methoxyphenyl)-5-oxopyrrolidine-3-carboxylic acid
Catalog # APOSOR9334-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:235.24 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:56617-47-1
Formula:C₁₂H₁₃NO₄
MDL Number:MFCD00227562