Order Entry
Czech Republic
ContactUsLinkComponent
(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione 97%
(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione 97%
Catalog # APOSOR933335-25G
Supplier:  Apollo Scientific
CAS Number:  
(3S)-(-)-cis-3,6-Dimethyl-1,4-dioxane-2,5-dione 97%
Catalog # APOSOR933335-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :144.13 g/mol
Storage Temperature:Ambient
CAS Number:4511-42-6
Formula:C₆H₈O₄
MDL Number:MFCD00070594