Order Entry
Czech Republic
ContactUsLinkComponent
meso-1,4-Dichloro-2,3-butanediol 95%
meso-1,4-Dichloro-2,3-butanediol 95%
Catalog # APOSOR933328-5G
Supplier:  Apollo Scientific
CAS Number:  
meso-1,4-Dichloro-2,3-butanediol 95%
Catalog # APOSOR933328-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :159.01 g/mol
Storage Temperature:Ambient
CAS Number:7268-35-1
Formula:C₄H₈Cl₂O₂