Order Entry
Czech Republic
ContactUsLinkComponent
C-[1,4]Dioxan-2-ylmethylamine 95%
C-[1,4]Dioxan-2-ylmethylamine 95%
Catalog # APOSOR932330-250MG
Supplier:  Apollo Scientific
CAS Number:  
C-[1,4]Dioxan-2-ylmethylamine 95%
Catalog # APOSOR932330-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :117.15 g/mol
Storage Temperature:Refrigerator
UN:3259
ADR:8,III
CAS Number:88277-83-2
Formula:C₅H₁₁NO₂
MDL Number:MFCD01994527