Order Entry
Czech Republic
ContactUsLinkComponent
1-(1H-Indol-2-yl)ethanone 95%
1-(1H-Indol-2-yl)ethanone 95%
Catalog # APOSOR931775-1G
Supplier:  Apollo Scientific
CAS Number:  
1-(1H-Indol-2-yl)ethanone 95%
Catalog # APOSOR931775-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :159.19 g/mol
Storage Temperature:Ambient
CAS Number:4264-35-1
Formula:C₁₀H₉NO
MDL Number:MFCD01863329