Order Entry
Czech Republic
ContactUsLinkComponent
 
1H-Pyrazolo[3,4-b]pyridine-3-carbonitrile 95%
Catalog # APOSOR931536-1G
Supplier:  Apollo Scientific
CAS Number:  
1H-Pyrazolo[3,4-b]pyridine-3-carbonitrile 95%
Catalog # APOSOR931536-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :144.14 g/mol
Storage Temperature:Ambient
CAS Number:956010-88-1
Formula:C₇H₄N₄
MDL Number:MFCD13176785